Learn More
Omega Engineering, Inc. 4-Channel Handheld Data Logger Thermometer
Catalog No. NC1616147
Encompass
The HH374 has a high resolution and fast analog to digital converter. Measurements settings and results are shown on the backlit LCD display. Data can be stored in the meter or directly saved on a computer through PC interface. Recorded data can be further processed on a computer. With the exclusive software, recording/recalling of data and programming of logging parameters can be done directly through a PC.
Catalog No. NC1616147 Supplier Omega Engineering, Inc. Supplier No. HH374
The HH374 has a high resolution and fast analog to digital converter. Measurements settings and results are shown on the backlit LCD display. Data can be stored in the meter or directly saved on a computer through PC interface. Recorded data can be further processed on a computer. With the exclusive software, recording/recalling of data and programming of logging parameters can be done directly through a PC.