Learn More
Invitrogen™ Hand-Held Magnetic Plate Washer
Catalog No. 502557501
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
Catalog No. Quantity
50-255-7501 1 Ea.
1 options
Catalog No. 50-255-7501 Supplier Invitrogen™ Supplier No. EPX55555000
Add to Cart
Edge
Add to Cart
The Hand-Held Magnetic Plate Washer is designed for use with the ProcartaPlex assay configured for magnetic separation.
The Hand-Held Magnetic Plate Washer is designed for use with the ProcartaPlex assay configured for magnetic separation. The handheld magnetic separator enables you to remove the liquid contents of the 96-well plate by simply decanting or flicking the contents into a sink and blotting off the remainder on a paper towel.
Please refer to the appropriate ProcartaPlex User Manual for details on running the assay.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Plate Washer
Quantity 1 Ea.
For Research Use Only. Not for use in diagnostic procedures.