Learn More
Sigma Aldrich Fine Chemicals Biosciences Hand operated aluminum cap1EA
Catalog No. 502905833
Encompass
Used to attach aluminum seals to bottles and vials with a crimp/serum finishCushioned ergonomic handle reduces hand fatigueLabeled for quick size identificationPolished crimping jaws provide consistent sealing2 year warranty
Catalog No. 50-290-5833 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1142431EA
Used to attach aluminum seals to bottles and vials with a crimp/serum finishCushioned ergonomic handle reduces hand fatigueLabeled for quick size identificationPolished crimping jaws provide consistent sealing2 year warranty