Learn More
Electron Microscopy Sciences Hand Piece, Procraft
SDP
Catalog No. 50996802
Encompass_Preferred
The most popular hand pieces for use. The chuck takes accessory shanks from 0 to 5/32" (0 to 4 mm). Includes chuck key.
Catalog No. 50-996-802 Supplier Electron Microscopy Sciences Supplier No. 6213701
The most popular hand pieces for use. The chuck takes accessory shanks from 0 to 5/32" (0 to 4 mm). Includes chuck key.