Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hand Pointers
SCI_ED

Catalog No. p-3592362 Shop All Learning Resources, Inc Products
Change view
Click to view available options
Length (English):
12 in.
24 in.
Quantity:
3/Pk.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Quantity
S95842 24 in. 3/Pk.
S95843 12 in. 3/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Catalog No. S95842 Supplier Learning Resources, Inc Supplier No. LER1960
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHOKING HAZARD —Small parts. Not for children under 3 yrs.

Encourage attention and participation.

  • Draws attention to interactive whiteboards, charts, maps, and more
  • Reinforces fluency, vocabulary, and phonics as you point to parts of a text, pocket chart, or big book
  • Available in 11, 12 or 24 in. lengths (27.9, 30.5, 61cm)
  • 24 in. Pointers come with convenient metal hanger

Order Info

Grades PreK+.

Specifications

Grade Level Early Childhood, Elementary, Intermediate - Middle School, High School
Grade Pre-K to 12
For Use With (Equipment) Whiteboards, charts, maps and more
Product Type Hand Pointers
Includes One of each red, green and blue, metal hangers
Quantity 3/Pk.
For Use With (Application) Encourages attention and participation
Subject Safety and Training Materials
Length (English) 24 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.