Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Walter Stern Hand-Powered Blower

Catalog No. 02680 Shop All Walter Stern Inc. Products
Change view
Click to view available options
For Use With:
Blowpipes
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
02-680 Blowpipes
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-680 Supplier Walter Stern Supplier No. 206
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Provides continuous stream of air at low pressure

  • Consists of tube bulb and 20 in. (51cm) outlet tube
  • Net-covered second bulb serves as reservoir
  • For use with blowpipes and other small apparatus
  • Made of latex rubber

Specifications

For Use With Blowpipes
Includes Tube bulb and 20 in. outlet tube
Overall Length (English) 28 in.
Overall Length (Metric) 71.12 cm
Type Blower
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.