Learn More
Electron Microscopy Sciences Paraffin Tape-Transfer System Hand Roller
SDP
Catalog No. 5030377
Encompass
The Paraffin Tape-Transfer System is not available. Kit components are available for purchase. Required in the CryoJane process for laminating Tape Window to block-face, use with Paraffin Sectioning Tape Transfer System.
Catalog No. 50-303-77 Supplier Electron Microscopy Sciences Supplier No. 6280046
The Paraffin Tape-Transfer System is not available. Kit components are available for purchase. Required in the CryoJane process for laminating Tape Window to block-face, use with Paraffin Sectioning Tape Transfer System.