Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ricca Chemical Hand Sanitizer
SDP

Catalog No. R421900020F Shop All Ricca Chemical Company Products
Change view
Click to view available options
Volume (Metric):
20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
R421900020F 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. R421900020F Supplier Ricca Chemical Supplier No. R421900020F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ricca Chemical Hand Sanitizer is formulated to meet WHO guidelines and FDA requirements for effectiveness. WHO formula II.

  • Active ingredient: 75% Isopropyl Alcohol.
  • Formulated to meet World Health Organization (WHO) and FDA requirements for effectiveness.
  • Manufactured in an ISO 9001:2015 accredited, cGMP, and FDA registered facility.
  • FDA compliant labelling, safety data sheet and certificate of analysis available 24/7 on the website.

Specifications

Product Type Hand Sterilizer/Disinfectant
Color Colorless
Form Gel
Packaging Cubitainer™
Volume (English) 5.3 gal.
Volume (Metric) 20 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.