Learn More
Grainger Hand Sanitizer Alcohol Free 1.5oz PK24
Catalog No. 502764300
Encompass_Preferred
The Kimberly-Clark Professional KCP100055CT Hand Sanitizer is an alcohol-based hand sanitizer gel that kills 99.9% of most common germs. It contains 62% ethyl alcohol and is formulated with moisturizers to leave hands feeling soft. This 1 liter bottle with pump dispenser provides waterless hand hygiene when soap and water are unavailable. It can help reduce bacteria on hands and prevent cross-contamination. The gel is fragrance-free and dye-free.
Catalog No. 50-276-4300 Supplier Grainger Supplier No. 170K55
The Kimberly-Clark Professional KCP100055CT Hand Sanitizer is an alcohol-based hand sanitizer gel that kills 99.9% of most common germs. It contains 62% ethyl alcohol and is formulated with moisturizers to leave hands feeling soft. This 1 liter bottle with pump dispenser provides waterless hand hygiene when soap and water are unavailable. It can help reduce bacteria on hands and prevent cross-contamination. The gel is fragrance-free and dye-free.