Learn More
AeroCleanse Hand Sanitizer with Pump Top
Description
- Reduce disease-causing bacteria when soap and water are not available
- Pump top
- Available in Antiseptic Gel or Liquid
Specifications
Specifications
| Product Type | Hand Sanitizer |
| Form | Gel |
| Composition | 80% Alcohol Antiseptic Liquid |
| Volume (English) | 1 gal. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.