Learn More
Description
- Cruelty Free
- Silicone and Paraben Free
- Synthetic Fragrance Free
Specifications
Specifications
| Product Type | Hand Sanitizer |
| Color | Clear |
| Form | Spray |
| Certifications/Compliance | FDA Approved Formula |
| For Use With (Application) | For external/topical use only |
| Packaging | Spray Bottle |
| Quantity | 12/Cs. |
| Volume (English) | 4 oz. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.