Learn More
Lakeside Manufacturing Inc Hand Washing Station Cold Only
Supplier:  Lakeside Manufacturing Inc 9600
Encompass_Preferred
Hand Washing Stations by Lakeside Manufacturing provide worker hand sanitation in healthcare environments with compact and self-contained functionality. The Lakeside 9600 Hand Washing Station with (1) 10" x 12" x 5" sink bowl and cold faucet has a 5 gallon fresh water capacity for approx. 35 hand washings. Built-in fresh water supply for sanitary handwashing and waste water storage. Sink and cabinet are constructed of durable, easy to clean and sanitize stainless steel. Heavy-duty chrome plated brass faucet has easy-to-use on/off paddle handle. Chrome plated soap dispenser is securely mounted next to the sink. Locking door latch prevents unauthorized access to the interior. 5" no-mark cushion-tread wheels, 2 swivel w/ brake and 2 fixed, for easy navigation. Waste water capacity is 7 gallons. Pump features automatic water demand - no priming, smooth water flow, backflow protection - no fresh water contamination, 150 PSI operating pressure and overheat cut-off protection. Made in USA.
Catalog No. 50-288-4443
Add to Cart
Request a Quote