Learn More
Lakeside Manufacturing Inc Hand Washing Station Hot-Cold
Supplier:  Lakeside Manufacturing Inc 9610
Encompass_Preferred
The Lakeside 9610 Hand Washing Station with (1) 10" x 12" x 5" sink bowl and hot and cold faucet has a 10 gallon fresh water capacity for approximately 70 hand washings. Built-in fresh water supply for sanitary handwashing and waste water storage. Sink and cabinet are constructed of durable, easy to clean and sanitize stainless steel. Heavy-duty chrome plated brass faucet has easy-to-use on/off paddle handle. Chrome plated soap dispenser is securely mounted next to the sink. Locking door latch prevents unauthorized access to the interior. 5" no-mark cushion-tread wheels, 2 swivel w/ brake and 2 fixed, for easy navigation. Includes hot running water and twice the water and waste capacity. Hot water heater has 2-1/2 gallon capacity and heats in 15 minutes for efficient service. Water heater features adjustable thermostat (110­­°F - 170°F), replaceable heating element, corrosion-resistant bronze tank, 150 PSI operating pressure and on/off switch on unit. Made in the USA.
Catalog No. 50-288-4487
Add to Cart
Request a Quote