Learn More
Lakeside Manufacturing Inc Hand Washing Station Hot Only
Catalog No. 502884321
Encompass_Preferred
Hand Washing Stations by Lakeside Manufacturing provide worker hand sanitation in healthcare environments with compact and self-contained functionality. The Lakeside 9620 Hand Washing Station with (1) 10" x 12" x 5" sink bowl and hot faucet has a 5 gallon fresh water capacity for approximately 35 hand washings. Built-in fresh water supply for sanitary handwashing and waste water storage. Sink and cabinet are constructed of durable, easy to clean and sanitize stainless steel. Heavy-duty chrome plated brass faucet has easy-to-use on/off paddle handle. Chrome plated soap dispenser is securely mounted next to the sink. Locking door latch prevents unauthorized access to the interior. 5" no-mark cushion-tread wheels, 2 swivel w/ brake and 2 fixed, for easy navigation. Waste water capacity is 7 gallons. Water heater features adjustable thermostat (110­­°F - 170°F), replaceable heating element, corrosion-resistant bronze tank, 150 PSI operating pressure and on/off switch on unit. Made in USA.
Catalog No. 50-288-4321 Supplier Lakeside Manufacturing Inc Supplier No. 9620
Add to Cart
Add to Cart
Request a Quote
Hand Washing Stations by Lakeside Manufacturing provide worker hand sanitation in healthcare environments with compact and self-contained functionality. The Lakeside 9620 Hand Washing Station with (1) 10" x 12" x 5" sink bowl and hot faucet has a 5 gallon fresh water capacity for approximately 35 hand washings. Built-in fresh water supply for sanitary handwashing and waste water storage. Sink and cabinet are constructed of durable, easy to clean and sanitize stainless steel. Heavy-duty chrome plated brass faucet has easy-to-use on/off paddle handle. Chrome plated soap dispenser is securely mounted next to the sink. Locking door latch prevents unauthorized access to the interior. 5" no-mark cushion-tread wheels, 2 swivel w/ brake and 2 fixed, for easy navigation. Waste water capacity is 7 gallons. Water heater features adjustable thermostat (110­­°F - 170°F), replaceable heating element, corrosion-resistant bronze tank, 150 PSI operating pressure and on/off switch on unit. Made in USA.