Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Handaphos

Catalog No. 79958025MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-958-025MG 100 mg
79-958-0100MG 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 79-958-025MG Supplier Sigma Aldrich Supplier No. 79958025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.HandaPhos forms a 1:1 complex with Pd(OAc)2 thereby arriving at a pre-catalyst, used at the pp mLevel, that enables Suzuki-Miyaura cross-couplings to be run under micellar catalysis in water at room temperature. To be used in tandem with Designer Surfactant TPGS-750-M.Watch Professor Lipshutz talk about HandaPhos in this webinar:From milligrams to kilograms: synthetic chemistry following nature's lead

Specifications

CAS 88-99-3
Molecular Formula C35H47O3P
Quantity 100 mg
Molecular Weight (g/mol) 546.72 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.