Learn More
Hach Company Handbook, Safety
Supplier:  Hach Company 2239800
Encompass
Comprehensive guide uses a convenient tabular format to present key industrial hygiene and medical surveillance information on 397 chemicals and compounds. Topics: Chemical and physical properties, Permissible Exposure Limits (PEL), Immediately Dangerous to Life or Health (IDLH) levels, health hazards and chemical incompatibilities, personal protection recommendations and first aid procedures. Updated every four years.
Catalog No. 50-199-5597