Learn More
Bio Rad Laboratories HANDCASTING KIT WIDE MINI
Catalog No. 509011336
Encompass
Kit, Wide Mini Handcasting; Bio-Rad; Includes 15 x 7cm tray, casting gates, 15-well 1.5mm fixed-height comb, 20-well 1.5mm fixed-height comb
Catalog No. 50-901-1336 Supplier Bio Rad Laboratories Supplier No. 1704497
Kit, Wide Mini Handcasting; Bio-Rad; Includes 15 x 7cm tray, casting gates, 15-well 1.5mm fixed-height comb, 20-well 1.5mm fixed-height comb