Learn More
Medchemexpress LLC HY-N2083 5mg Medchemexpress, Handelin CAS:62687-22-3 Purity:>98%
SDP
Catalog No. 501970365
Encompass_Preferred
Medchemexpress, HY-N2083 5mg Handelin CAS:62687-22-3 Handelin is a guaianolide dimer from Chrysanthemum boreale that has potent anti-inflammatory activity by down-regulating NF-κB signaling and pro-inflammatory cytokine production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0365 Supplier Medchemexpress LLC Supplier No. HYN20835MG
Medchemexpress, HY-N2083 5mg Handelin CAS:62687-22-3 Handelin is a guaianolide dimer from Chrysanthemum boreale that has potent anti-inflammatory activity by down-regulating NF-κB signaling and pro-inflammatory cytokine production. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.