Learn More
Thomas Scientific 8-Channel Plastic Adapter for disposable tips
SDP
Catalog No. 501461444
Encompass
A Complete Set of Tools for Aspiration Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes! This ergonomic, hand-held aspirator can be used with Pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques. Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.
Catalog No. 50-146-1444 Supplier Thomas Scientific Supplier No. 1221X85
A Complete Set of Tools for Aspiration Rapidly remove supernatant from a variety of cell culture or multi-well plates and dishes! This ergonomic, hand-held aspirator can be used with Pasteur pipettes, plastic aspiration pipettes or a variety of adapters for specific applications. A pressure-sensitive button on the top of the unit controls the specific amount of vacuum applied. This prevents the accidental aspiration of cells or supernatant associated with normal vacuum techniques. Use with standard in-house vacuum source or with the M-Vac or E-Vac aspiration systems. Autoclavable unit is supplied with a single-channel adapter for disposable pipette tips.