Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Handheld Analog Clinical Refractometer
GREENER_CHOICESDP

Catalog No. 12561341 Shop All Fisherbrand Products
Change view
Click to view available options
Temperature Compensation:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Compensation
12-561-341 Yes
12-561-340 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-561-341
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Calibration Screw Driver, Soft sided case, Applicator, Manual

When you need reliable, accurate readings of your Urine samples, the Fisherbrand™ Analog Clinical Refractometers delivers the value with the ability to read Serum Protein, Specific Gravity and Refractive Index all on a single scale.

Refractometers have been used for measuring the concentration of solutions for more than a century. Their primary purpose is to measure the degree at which light bends as it passes through a medium, and is used in a wide range of applications and industries as a quality control and diagnostic tool.

Fisherbrand Analog Refractometers are designed for simple operation and built with an all-metal construction for durability. Its industrial design is the most commonly used design among refractometer manufactures as it is a cost-effective model to build.

Each unit is inspected in the United States by a factory qualified technician under a rigid quality control process to ensure its accuracy, and performance.

Key Features:

  • Quickly calibrate the instrument with a turn of a screw, and distilled water
  • Comfortable eyecup blocks out ambient light and allows for crystal clear viewing of readings
  • Automatic temperature compensation prevents the need for constant re-calibration of the instrument. As the ambient temperature changes, the scale will automatically compensate for the difference in the degree of refraction. This ensures an accurate reading every time
  • All metal construction creates durability. NOTE: Model 12-561-340 comes without ATC
  • Footrest allows the instrument to be set on a flat surface without the worry of it rolling off working surfaces
  • Rubber hand grip is comfortable, easy to grasp. Its non-slip pattern grip reduces that chance of accidentally dropping the instrument
  • Double hinged daylight plates create two points of movement to allow for greater flexibility in movement; making it easy to add samples and clean
  • Portable, easy to use design.

Clinical Models:

Quickly and accurately get an indication of basic health parameters such as dehydration and blood plasma protein levels. Fisherbrand Analog Clinical Refractometers use three-scales to measure Total Serum Protein, Urine Specific Gravity, and Refractive Index. They are ideal for use in quality control, research, and clinical laboratories.

  • Urine Specific Gravity: 1.000ppt to 1.050ppt with granularity to measure down to 0.001ppt at a 0.002ppt degree of accuracy
  • Serum Protein: 1g to 12g with granularity to measure down to 0.2g/100mL at a 0.2g/100mL degree of accuracy
  • Refractive Index: 1.334nD to 1.360nD with granularity to measure down to .0005nD at a 0.0005nD degree of accuracy.

Warranty and Services

2 years

Specifications

Type Clinical Refractometer
Scale Range 1.000 to 1.050 Urine Specific Gravity, 1 to 12 g/dl Serum Protein
Refractive Index 1.331 to 1.36
Sample Volume 1 mL
Calibration Distilled Water
Accuracy 0.002; 0.2 g/dl; 0.0005
Resolution 0.0005
Increments 0.002; 0.2 g/dl; 0.0005
Scale Type Linear
Lighting Type Ambient
For Use With (Application) Clinical
Includes Calibration Screw Driver, Soft sided case, Applicator, Manual
IP Rating IP54
Quantity 1
Weight (English) 0.881 lb.
Weight (Metric) 0.4 kg
Dimensions (L x W x H) 172 x 39 x 39 mm
Height (English) 1.535 in.
Height (Metric) 39 mm
Length (English) 6.772 in.
Length (Metric) 204 mm
Width (English) 1.535 in.
Width (Metric) 39 mm
Temperature Compensation Yes
Temperature Range (Metric) 10°C to 30°C
Warranty 5 years optical and mechanical, 1 year electrical from date of purchase
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.