Learn More
MSC EMist Compact Electrostatic Disinfectant Sprayer, Cordless, 8 oz, 1 Kit
Catalog No. 502866232
Encompass
The EMist EPIX360 Compact Electrostatic Disinfectant Sprayer is a cordless, handheld device designed for efficient application of water-soluble disinfectants. It utilizes intelligent electrostatic technology to provide superior coverage for healthier spaces.
  • Intelligent electrostatic disinfection
  • Cordless operation for mobility
  • Has 8 oz tank capacity
  • Covers up to 4,000 high-touch point sq.ft.
  • Lightweight and compact design
  • Features bipolar polarity for improved transfer efficiency
  • Eliminates shock and electrical hazards
  • Includes sprayer, tank, 2 batteries, charger, carrying case, and training toolkit
Catalog No. 50-286-6232 Supplier MSC Supplier No. 10301398
The EMist EPIX360 Compact Electrostatic Disinfectant Sprayer is a cordless, handheld device designed for efficient application of water-soluble disinfectants. It utilizes intelligent electrostatic technology to provide superior coverage for healthier spaces.
  • Intelligent electrostatic disinfection
  • Cordless operation for mobility
  • Has 8 oz tank capacity
  • Covers up to 4,000 high-touch point sq.ft.
  • Lightweight and compact design
  • Features bipolar polarity for improved transfer efficiency
  • Eliminates shock and electrical hazards
  • Includes sprayer, tank, 2 batteries, charger, carrying case, and training toolkit