Learn More
Grainger Handheld Capacitance Meter LCD
Catalog No. 502753479
Encompass_Preferred
The BK Precision 810B Handheld Capacitance Meter is a portable instrument for measuring capacitance. It has a capacitance range of 0.1 pF to 200 μF and can measure capacitors, cables, batteries and more. The large LCD display shows the capacitance value and test conditions. Features include relative measurement, zero adjustment, and low battery indicator. It is powered by a 9V battery and comes with test leads, battery and manual. The compact handheld design makes it convenient for field service, production testing, and bench use.
Catalog No. 50-275-3479 Supplier Grainger Supplier No. 14H638
The BK Precision 810B Handheld Capacitance Meter is a portable instrument for measuring capacitance. It has a capacitance range of 0.1 pF to 200 μF and can measure capacitors, cables, batteries and more. The large LCD display shows the capacitance value and test conditions. Features include relative measurement, zero adjustment, and low battery indicator. It is powered by a 9V battery and comes with test leads, battery and manual. The compact handheld design makes it convenient for field service, production testing, and bench use.