Learn More
Grainger Handheld Flashlight Lexan Yellow 183lm
Catalog No. 502753262
Encompass_Preferred
The Pelican 183 Safety Rated Flashlight is a durable, waterproof flashlight designed for demanding work environments. It features a C4 LED bulb that produces 140 lumens of light on high mode and runs for 29 hours on low mode. The flashlight is impact resistant, chemical resistant, and rated for hazardous locations. It is made from a high-impact polymer housing with rubber overmolding and has an anti-slip grip. The flashlight measures 6.37" long x 1.37" diameter and weighs 7 oz. It requires 2 AA batteries, which are included. The 183 flashlight is UL classified for safety and carries a lifetime guarantee.
Catalog No. 50-275-3262 Supplier Grainger Supplier No. 14L589
The Pelican 183 Safety Rated Flashlight is a durable, waterproof flashlight designed for demanding work environments. It features a C4 LED bulb that produces 140 lumens of light on high mode and runs for 29 hours on low mode. The flashlight is impact resistant, chemical resistant, and rated for hazardous locations. It is made from a high-impact polymer housing with rubber overmolding and has an anti-slip grip. The flashlight measures 6.37" long x 1.37" diameter and weighs 7 oz. It requires 2 AA batteries, which are included. The 183 flashlight is UL classified for safety and carries a lifetime guarantee.