Learn More
Grainger Handheld Sprayer 2 gal 4 3/16 ft L
Catalog No. 502749726
Encompass_Preferred
The Grainger 14U075 Handheld Sprayer Spot is a handheld sprayer designed for spot spraying applications. It features a translucent polyethylene tank that holds 1 gallon of liquid and has measurement markings for easy filling and mixing. The wand is made of polypropylene with a Viton seal and can spray up to 30 psi with an adjustable nozzle. It has a comfort grip handle and includes a 36 inch hose. This handheld sprayer is ideal for applying pesticides, herbicides, fertilizers, cleaners and other liquids in home, farm, commercial and industrial settings. It provides targeted application for small jobs.
Catalog No. 50-274-9726 Supplier Grainger Supplier No. 14U075
The Grainger 14U075 Handheld Sprayer Spot is a handheld sprayer designed for spot spraying applications. It features a translucent polyethylene tank that holds 1 gallon of liquid and has measurement markings for easy filling and mixing. The wand is made of polypropylene with a Viton seal and can spray up to 30 psi with an adjustable nozzle. It has a comfort grip handle and includes a 36 inch hose. This handheld sprayer is ideal for applying pesticides, herbicides, fertilizers, cleaners and other liquids in home, farm, commercial and industrial settings. It provides targeted application for small jobs.