Learn More
Grainger Handheld Sprayer 2 gal 4 ft L
Catalog No. 502765054
Encompass_Preferred
The SOLO Handheld Sprayer Spot is a handheld sprayer designed for spot spraying weeds and other unwanted vegetation. It features a 1 gallon translucent tank for monitoring fluid levels and has a pump handle that builds up to 90 PSI of pressure for consistent spraying power. The adjustable brass cone nozzle provides a variable spray pattern from a fine mist to a solid stream. It has a comfort grip handle and includes a shoulder strap for easy carrying during use. The translucent tank allows you to easily monitor fluid levels. Overall, this is a versatile handheld sprayer suitable for spot spraying unwanted vegetation in your yard or garden.
Catalog No. 50-276-5054 Supplier Grainger Supplier No. 16T951
The SOLO Handheld Sprayer Spot is a handheld sprayer designed for spot spraying weeds and other unwanted vegetation. It features a 1 gallon translucent tank for monitoring fluid levels and has a pump handle that builds up to 90 PSI of pressure for consistent spraying power. The adjustable brass cone nozzle provides a variable spray pattern from a fine mist to a solid stream. It has a comfort grip handle and includes a shoulder strap for easy carrying during use. The translucent tank allows you to easily monitor fluid levels. Overall, this is a versatile handheld sprayer suitable for spot spraying unwanted vegetation in your yard or garden.