Learn More
Grainger Handheld Sprayer 3 gal 4 3/16 ft L
Catalog No. 502749727
Encompass_Preferred
The Grainger 14U076 Handheld Sprayer Spot is a handheld sprayer designed for spot spraying weeds and applying pesticides or fertilizers in a targeted manner. It features a translucent tank that holds 1 gallon of liquid and is equipped with Viton seals for chemical resistance. The wand is made of polypropylene with an adjustable brass cone nozzle and the pump handle provides consistent pressure up to 60 PSI. This handheld sprayer is lightweight at just 2.2 lbs when empty and has a comfortable pistol grip handle and shoulder strap for easy carrying and operation. It's ideal for small spraying jobs in home gardens, landscaping applications, and agricultural use.
Catalog No. 50-274-9727 Supplier Grainger Supplier No. 14U076
The Grainger 14U076 Handheld Sprayer Spot is a handheld sprayer designed for spot spraying weeds and applying pesticides or fertilizers in a targeted manner. It features a translucent tank that holds 1 gallon of liquid and is equipped with Viton seals for chemical resistance. The wand is made of polypropylene with an adjustable brass cone nozzle and the pump handle provides consistent pressure up to 60 PSI. This handheld sprayer is lightweight at just 2.2 lbs when empty and has a comfortable pistol grip handle and shoulder strap for easy carrying and operation. It's ideal for small spraying jobs in home gardens, landscaping applications, and agricultural use.