Learn More
Omega Engineering, Inc. PT100 RTD Handheld Thermometer
Catalog No. NC1060260
Encompass
The HH370 is a waterproof, highly accurate handheld RTD temperature meter that uses the dedicated probes RTD-370-PT100 and RTD-370-PT1K. These probes have a water-resistant connection and a measurement range of -100 to 300°C (-148 to 572°F). The 100 omega probe is included with the unit.
Catalog No. NC1060260 Supplier Omega Engineering, Inc. Supplier No. HH370
The HH370 is a waterproof, highly accurate handheld RTD temperature meter that uses the dedicated probes RTD-370-PT100 and RTD-370-PT1K. These probes have a water-resistant connection and a measurement range of -100 to 300°C (-148 to 572°F). The 100 omega probe is included with the unit.