Learn More
FLOW CLEANROOMS & SUPPLY H303 Handheld Airborne Particle Counter, Size Range: Measures 0.3 to 25.0 µ, Flow Rate: 0.1 CFM (2.83 LPM),
Catalog No. NC2953954
Encompass
This Airy Technology Handheld Particle Counter offers the industry's highest concentration of 15,000,000 particles/ft³ at 10% coincidence loss. The H303 model is ideal for use in cleanrooms, industrial health & safety, and air quality applications. One of the most economical devices with a color display that displays up to 3 user-selectable size channels as well as temperature and relative humidity. You can view data on screen or export to Instrument Management Software (IMS). Airy Technology counters can be controlled and monitored remotely via a web browser.
Catalog No. NC2953954 Supplier FLOW CLEANROOMS & SUPPLY Supplier No. 7201006
This Airy Technology Handheld Particle Counter offers the industry's highest concentration of 15,000,000 particles/ft³ at 10% coincidence loss. The H303 model is ideal for use in cleanrooms, industrial health & safety, and air quality applications. One of the most economical devices with a color display that displays up to 3 user-selectable size channels as well as temperature and relative humidity. You can view data on screen or export to Instrument Management Software (IMS). Airy Technology counters can be controlled and monitored remotely via a web browser.