Learn More
ANGSTROM INC Handy-Kaps. 1000/pk
Catalog No. NC1646368
Encompass
Angstrom Handy-Kaps are thin-walled aluminum sample cups that are useful for preparing many types of samples prior to XRF or OE analysis. Cups are loaded with powder, then pressed to produce a reinforced sample that can be easily marked for sample identification.
Catalog No. NC1646368 Supplier ANGSTROM INC Supplier No. 2800
Angstrom Handy-Kaps are thin-walled aluminum sample cups that are useful for preparing many types of samples prior to XRF or OE analysis. Cups are loaded with powder, then pressed to produce a reinforced sample that can be easily marked for sample identification.