Learn More
Waters Corp Finger-Tight Fitting, Polypropylene, Clear, 1/16 in. Tubing, 10-pack
Catalog No. 50819095 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These finger-tight fittings are made from polypropylene, providing a clear design for easy visual inspection. They are supplied in a convenient pack of 10 and are suitable for 1/16 in. tubing.
  • Made from polypropylene material
  • Clear color for visibility
  • Supplied in a 10-pack
  • Designed for 1/16 in. tubing
Catalog No. 50-819-095 Supplier Waters Corp Supplier No. PSL618022
These finger-tight fittings are made from polypropylene, providing a clear design for easy visual inspection. They are supplied in a convenient pack of 10 and are suitable for 1/16 in. tubing.
  • Made from polypropylene material
  • Clear color for visibility
  • Supplied in a 10-pack
  • Designed for 1/16 in. tubing