Learn More
Waters Corp Handilok Fitting, CTFE, 1/16 in. Tubing, Tool-Free
Catalog No. 50793436 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Handilok fitting is designed for reliable connections in laboratory systems. It features durable CTFE construction and tool-free installation for convenience, making it suitable for applications requiring 1/16 in. tubing.
  • CTFE construction for chemical resistance and durability.
  • Tool-free installation for quick and easy setup.
  • Designed for 1/16 in. tubing.
Catalog No. 50-793-436 Supplier Waters Corp Supplier No. PSL618021
This Handilok fitting is designed for reliable connections in laboratory systems. It features durable CTFE construction and tool-free installation for convenience, making it suitable for applications requiring 1/16 in. tubing.
  • CTFE construction for chemical resistance and durability.
  • Tool-free installation for quick and easy setup.
  • Designed for 1/16 in. tubing.