Learn More
Electron Microscopy Sciences Pin Vise Needle and Knife Interchangeable Handle, 92 mm OAL
SDP
Catalog No. 5032141
Encompass_Preferred
Made from stainless steel, pin-vise type Handle, which accepts either dissecting needles or a knife. The handle is 92 mm L x 6 mm diameter. 92mm (3-5/8") long x 6mm (1/4") diameter.
OAL: 92 mm
Catalog No. 50-321-41 Supplier Electron Microscopy Sciences Supplier No. 72953HD
Made from stainless steel, pin-vise type Handle, which accepts either dissecting needles or a knife. The handle is 92 mm L x 6 mm diameter. 92mm (3-5/8") long x 6mm (1/4") diameter.
OAL: 92 mm