Learn More
World Precision Instrument Titanium Handle for Angled Sapphire Blades
SDP
Catalog No. 50822837
Encompass
The Titanium Scalpel Handle is for use with our Sapphire Blades. The 501811 version is for use with the regular sapphire blades (504074, 504075, and 504168), and the 501812 version is for use with angled sapphire blades. Since sapphires are one of the hardest materials available (second only to diamonds), they make super sharp blades for precise, clean cuts. They are non-magnetic, especially when combined with a titanium handle. Sapphire blades are non-conductive, which may be important if you are using electrical instruments. Sapphire blades are impervious to corrosion and extremely durable. The sapphire blades are sold separately. These handles are retractable, so you can protect the blade when it is not in use. The reusable scalpel handle is 13 cm (5.1") long.  They are constructed of titanium and are autoclavable or suitable for chemical sterilization.
Catalog No. 50-822-837 Supplier World Precision Instrument Supplier No. 501812
The Titanium Scalpel Handle is for use with our Sapphire Blades. The 501811 version is for use with the regular sapphire blades (504074, 504075, and 504168), and the 501812 version is for use with angled sapphire blades. Since sapphires are one of the hardest materials available (second only to diamonds), they make super sharp blades for precise, clean cuts. They are non-magnetic, especially when combined with a titanium handle. Sapphire blades are non-conductive, which may be important if you are using electrical instruments. Sapphire blades are impervious to corrosion and extremely durable. The sapphire blades are sold separately. These handles are retractable, so you can protect the blade when it is not in use. The reusable scalpel handle is 13 cm (5.1") long.  They are constructed of titanium and are autoclavable or suitable for chemical sterilization.