Learn More
Med Vet International Miltex Knife Handle, 4-7/8in, #3, Holds Blades 10 through 15C, MH4-7
Supplier:  Med Vet International MILMH47
Encompass_Preferred
Miltex Knife Handle, 4-7/8in, #3, Holds Blades 10 through 15C, MH4-7
Catalog No. 50-264-3488