Learn More
Med Vet International Miltex Knife Handle, 5-1/4in, #4, Holds Blades 20 Through 25, Mh4-8
Catalog No. 502831238
Encompass_Preferred
Miltex Knife Handle, 5-1/4in, #4, Holds Blades 20 through 25, MH4-8
Catalog No. 50-283-1238 Supplier Med Vet International Supplier No. MILMH48
 Restricted Item
Add to Cart
Add to Cart
Miltex Knife Handle, 5-1/4in, #4, Holds Blades 20 through 25, MH4-8