Learn More
Med Vet International #7 Knife Handle, for Blades 10-15c, 6.5", German Grade, Von Klaus
Supplier:  Med Vet International VK1012707
Encompass_Preferred
#7 German Grade knife handle can fit blades #10, #11, #12, and #15c surgical blades. This knife handle is used to create fine, precise incisions made in small spaces or deep wounds. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-304-5627