Learn More
Med Vet International Dynarex Yankauer Suction Handle, Bulb Tip, Non-Vented, w/Pre-Attached 1/4" x 6' Suction, 50/Case
Catalog No. 503045845
Encompass_Preferred
The Dynarex Resp-02™ Yankauer Suction Handle is designed to safely and effectively remove particulates and fluids from patients’ airways. The curved, one-piece Handle is constructed from transparent materials for clear visualization and features a slip-resistant, ribbed design to ensure a secure connection to suction tubing. Designed to clear airways to help improve ventilation and oxygenation Constructed from shatter-resistant plastic materials Available in vented, non-vented, bulb-tip, and straight-tip designs Sterile, single-use, and disposable for optimal hygiene Not made with natural rubber latex or DEHP
Catalog No. 50-304-5845 Supplier Med Vet International Supplier No. 32126
The Dynarex Resp-02™ Yankauer Suction Handle is designed to safely and effectively remove particulates and fluids from patients’ airways. The curved, one-piece Handle is constructed from transparent materials for clear visualization and features a slip-resistant, ribbed design to ensure a secure connection to suction tubing. Designed to clear airways to help improve ventilation and oxygenation Constructed from shatter-resistant plastic materials Available in vented, non-vented, bulb-tip, and straight-tip designs Sterile, single-use, and disposable for optimal hygiene Not made with natural rubber latex or DEHP