Learn More
Electron Microscopy Sciences Swann-Morton Blade Handle Size 7
SDP
Catalog No. 501928496
Encompass_Preferred
The high quality handes for surgical blades are made of are stainless steel.
Fits: Blades 6 - 16
Catalog No. 50-192-8496 Supplier Electron Microscopy Sciences Supplier No. 72054101
The high quality handes for surgical blades are made of are stainless steel.
Fits: Blades 6 - 16