Learn More
Med Vet International #3 Knife Handle, Fits Blades 10-15c, No Scale, Von Klaus German Surgical Steel
Catalog No. 502092262
Encompass_Preferred
#3 German Grade knife handle fit blades #10, #11, #12, and #15c surgical blades. These knife handles are commonly used for dissecting or making incisions. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2262 Supplier Med Vet International Supplier No. VK1012703
#3 German Grade knife handle fit blades #10, #11, #12, and #15c surgical blades. These knife handles are commonly used for dissecting or making incisions. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.