Learn More
Instech Handling tool for magnetic mouse Vascular Access Buttons
SDP
Catalog No. NC1265533
Encompass
The Instech VABMG handling tool is designed to securely hold magnetic mouse Vascular Access Buttons™ and the mouse itself without the need for hemostats. Its magnetic interface stabilizes the button during catheter access, sampling, or tether connection, improving safety and handling efficiency. This tool helps prevent damage to the VAB™ and minimizes stress on the animal when positioning or connecting accessories.
Features
  • Handling tool for magnetic mouse VABs™
  • Holds button and mouse magnetically
  • Eliminates need for hemostats
  • Improves handling safety and precision
  • Protects VAB™ from mechanical damage
Catalog No. NC1265533 Supplier Instech Supplier No. VABMG
The Instech VABMG handling tool is designed to securely hold magnetic mouse Vascular Access Buttons™ and the mouse itself without the need for hemostats. Its magnetic interface stabilizes the button during catheter access, sampling, or tether connection, improving safety and handling efficiency. This tool helps prevent damage to the VAB™ and minimizes stress on the animal when positioning or connecting accessories.
Features
  • Handling tool for magnetic mouse VABs™
  • Holds button and mouse magnetically
  • Eliminates need for hemostats
  • Improves handling safety and precision
  • Protects VAB™ from mechanical damage