Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International™ HandPRO™ 7900 Series Controlled Environment Nitrile Gloves
BIOPHARMA

Catalog No. 17900933 Shop All Hourglass International Inc Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-900-933 X-Small
17-900-934 Small
17-900-935 Medium
17-900-936 Large
17-900-937 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 17-900-933 Supplier Hourglass International™ Supplier No. 7900
Add to Cart
Add to Cart
This item is not returnable. View return policy

Controlled environment gloves are made from patented LOW DERMA™ accelerator-free, sulfur-free, and ultra-low surfactant nitrile with enhanced strength and a secure wet grip. Double-bagged and suitable for ISO 6 or higher cleanrooms.

  • Accelerator-free nitrile for reduced risk of glove-related allergy
  • Not made with carbamates, thiurams or MBTs
  • Sulfur-free, static dissipative and safe for metal surfaces
  • Consistently low endotoxin levels
  • Low-dye for reduced risk of leaching
  • Powder free
  • Low-surfactant and finger-textured for a secure wet or dry grip

Specifications

Certifications/Compliance Compliant to ASTM and IEST standards, ASTM D412, ASTM D5151-99, ASTM D573
Certifications/Compliance (Europe) EN 420
Chlorinated No
Cleanroom Class Class 1000 (ISO 6)
Accelerator Free Yes
AQL 2.5
Color White
Disposable Single-use
Exterior Finish Partially Textured
Finger Thickness (English) 6.2 mil
Finger Thickness (Metric) 0.16 mm
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Palm Thickness (English) 4.3 mil
Palm Thickness (Metric) 0.11 mm
Powdered Powder Free
Product Type Controlled Environment Gloves
Size X-Small
Sterility Non-sterile
Thickness (English) 6.2 mil
Thickness (Metric) 0.16 mm
Manufacturing/QMS Audit Standards ISO 13485; ISO 9001
Model 7900
Packaging Type Double-Bagged
Series HandPRO™ 7900
Tensile Strength 29 MPa
Length (English) 12 in.
Length (Metric) 30 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.