Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hands-On Science Books
SCI_ED

Catalog No. p-2376343
Change view
Click to view available options
Product Type:
Our Solar System
Subject:
Physics
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Subject
S65933 Our Solar System Physics
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S65933 Supplier Walch Education Supplier No. 037632

Explore scientific concepts through memorable activities based in the context of everyday life.

  • Easily integrated into any science course
  • Correlate to the National Science Education Standards
  • Grades 7 to 9
  • Each book is 96 to 104 pages

ABCs of Chemistry

  • Measure heat from a candle, make soap and build a simple battery
  • Investigate factors affecting the speed of a reaction
  • Learn to separate mixtures
  • Plus 14 more activities

Atmosphere & Weather

  • The state we're in: Air is a gas!
  • What is weather? Lightning!
  • Convection: temperature differences and the motion of air, highs, lows and winds
  • Plus 14 more activities

Electricity & Magnetism

  • Generate static electricity
  • Explore magnetic fields, series circuits, conductors and insulators
  • Create electricity from ice water
  • Plus 11 more activities

Force & Motion

  • Making and adding vectors
  • Velocity and acceleration
  • Newton's 1st Law: Inertia
  • Newton's 2nd Law: Fnet = ma
  • Newton's 3rd Law: Action/reaction
  • Pressure is not force
  • Plus 11 more activities

Introduction to Biotechnology

  • Get familiar with basic concepts, techniques and issues in biotechnology
  • Making yogurt and kimchi
  • Learn about gel electrophoresis and running a gel
  • Investigate Building People and To Clone, or Not to Clone
  • Plus 14 more acitivities

Light & Color

  • What is the nature of light?
  • How can we separate pigments?
  • How do different materials affect the absorption of light?
  • How light affects light-sensitive chemicals
  • Plus 16 more activities

Nutrition, Mind & Body

  • Covers a variety of human anatomy and physiology topics such as:
    • Seeing can be deceiving
    • Can eating ice cream be painful?
    • What parts of the body are involved in maintaining balance?
    • How much water does a popcorn kernel contain?
    • Milk, hot dogs and nutritional math

Our Solar System

  • The scale of the solar system: How big are the planets?
  • How far apart are the planets?
  • The shape of planetary orbits
  • Plus 15 more activities

Plants

  • Which way will the roots grow?
  • Which color of light prevents chlorosis in geranium plants?
  • How glucose content of a potato affects its use
  • Plus 17 more activities

Rocks & Minerals

  • Crystal structure of salt
  • Mineral tests — hardness
  • Rock types and mineral identification
  • Weathering rates of rocks and minerals
  • Plus 11 more activities

Simple Machines

  • Investigate: What is force? What is work? What is power?
  • Mechanical advantages in a bicycle
  • How a wedge and pulley work
  • Plus 14 more activities

Water

  • How do ships float?
  • When can water carry an electric current?
  • How does water act like a lens?
  • Investigate life in a pond
  • Plus 16 more activities

Science Series

  • Earth Science Series – Includes one copy each of Atmosphere & Weather, Rocks & Minerals and Our Solar System.
  • Life Science Series – Includes one copy each of Arthropods, Nutrition, Mind & Body and Plants.
  • Physical Science Series – Includes one copy each of Chemistry, Electricity & Magnetism, Force & Motion, Light & Color, Simple Machines and Water.

Specifications

Grade 7 to 9
Format Book
Product Type Our Solar System
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.