Learn More
Contec™ Hands-Free Removal Tool and Bag Holder
GREENER_CHOICE
Catalog No. 19161027 Shop All Contec Inc. Products
Catalog No. 19-161-027 Supplier Contec™ Supplier No. 2940
Add to Cart
Add to Cart
A hands-free removal tool to use with the range of QuickTask Mop Heads.
  • This stainless steel tool simply hangs on the back of the cart and includes a frame for holding disposal bags
  • The waste bag holder can be used with sterile or non-sterile waste bags for complete hands free removal and disposal of dirty mop heads
  • In order to use this hands-free option, the QuickTask magnetic mop head frames must be used, 2906 (stainless steel) and 2699P (polypropylene)
Specifications
Autoclavable Autoclavable