Learn More
Ampco Handstyle Scraper
SDP
Catalog No. 19174256 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
19 in.
Catalog No. Length (English)
19-174-256 19 in.
1 options
Catalog No. 19-174-256 Supplier Ampco Supplier No. S1069
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant scraper. Made in the USA.
  • Scraper with a flat, spatula-like metal blade on one end and a yellow, translucent, fluted screwdriver-shaped handle on the other end.
  • Please see image for additional information regarding measurements.
  • Flat end width (a): 3 and 3/4 in.
  • Flat end length (b): 3 and 3/4 in.
  • Overall length: 19 in.
Specifications
Product Type Scraper
Length (English) 19 in.
Quantity 1 Ea.
Width (English) 3 -3/4 in.