Learn More
Med Vet International Hand Sanitizer Alcare Plus 9 oz. Ethyl Alcohol Foaming Aerosol Can, 639936, Each
Catalog No. 503044987
Encompass_Preferred
For use where immediate, waterless hand degerming compliance is needed Broad-spectrum antimicrobial activity against pathogenic microorganisms Provides hand antisepsis and moisturizing at the same time Mild formulation for hand comfort Formulated with emollients for high frequency professional use Use with dispenser T603Q7
Catalog No. 50-304-4987 Supplier Med Vet International Supplier No. 207082EA
For use where immediate, waterless hand degerming compliance is needed Broad-spectrum antimicrobial activity against pathogenic microorganisms Provides hand antisepsis and moisturizing at the same time Mild formulation for hand comfort Formulated with emollients for high frequency professional use Use with dispenser T603Q7