Learn More
Martor Secunorm Handy W/ Stainless Steel Blade
Catalog No. NC9338807
Encompass_Preferred
The Handy is a semi automatic retractable spring loaded safety knife for cutting general industrial materials.-Semi automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of stainless steel-Safer alternative to fixed blade utility knives for general cutting tasks
Catalog No. NC9338807 Supplier Martor Supplier No. 1444.02
The Handy is a semi automatic retractable spring loaded safety knife for cutting general industrial materials.-Semi automatic retractable blade for safety-Ergonomically friendly ambidextrous slider mechanism to engage the blade-Durable body made of stainless steel-Safer alternative to fixed blade utility knives for general cutting tasks