Learn More
Chemglass Life Sciences HandyStep Touch, Inclusive Universal AC Adapter and Shelf/Rack Mount
SDP
Catalog No. 501979440
Encompass_Preferred
HandyStep Touch, Inclusive Universal AC Adapter and Shelf/Rack Mount. Choose the HandyStep touch for your repetitive pipetting tasks - dispensing, auto-dispensing and the ability to store favorites. The HandyStep touch S adds such advanced features like multi-aspiration, sequential dispensing and titration. Paired with the new PD-Tip™ ll Dispenser Tips with tip size coding, they make the perfect team for easy pipetting.
Catalog No. 50-197-9440 Supplier Chemglass Life Sciences Supplier No. CLS3468200
HandyStep Touch, Inclusive Universal AC Adapter and Shelf/Rack Mount. Choose the HandyStep touch for your repetitive pipetting tasks - dispensing, auto-dispensing and the ability to store favorites. The HandyStep touch S adds such advanced features like multi-aspiration, sequential dispensing and titration. Paired with the new PD-Tip™ ll Dispenser Tips with tip size coding, they make the perfect team for easy pipetting.