Learn More
Chemglass Life Sciences HandyStep Touch / Inductive Charging Stand Bundle, Universal Power Supply
SDP
Catalog No. 501979441
Encompass_Preferred
HandyStep Touch / Inductive Charging Stand Bundle, Universal Power Supply. Choose the HandyStep touch for your repetitive pipetting tasks - dispensing, auto-dispensing and the ability to store favorites. The HandyStep touch S adds such advanced features like multi-aspiration, sequential dispensing and titration. Paired with the new PD-Tip™ ll Dispenser Tips with tip size coding, they make the perfect team for easy pipetting.
Catalog No. 50-197-9441 Supplier Chemglass Life Sciences Supplier No. CLS3468200P
HandyStep Touch / Inductive Charging Stand Bundle, Universal Power Supply. Choose the HandyStep touch for your repetitive pipetting tasks - dispensing, auto-dispensing and the ability to store favorites. The HandyStep touch S adds such advanced features like multi-aspiration, sequential dispensing and titration. Paired with the new PD-Tip™ ll Dispenser Tips with tip size coding, they make the perfect team for easy pipetting.