Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ HandyStrap™ Series N100 Particulate Respirators
SDP

Catalog No. 19047828 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Medium/Large
Quantity:
5/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size Quantity
19-047-828 Medium/Large 5/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-047-828 Supplier Moldex™ Supplier No. 2730N100

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

N100 disposable respirators feature a unique HandyStrap™ with buckle, allowing the mask to hang down around the user’s neck when not in use.

  • Cloth strap is cool and comfortable against the skin
  • HandyStrap™ with buckle, allows mask to hang down around the user’s neck when not in use preventing premature disposal
  • Dura-Mesh™ provides bi-directional protection from dirt on the outside and moisture on the inside
  • Soft foam nose cushion for added comfort and no pressure points
  • Softspun™ lining for increased comfort and durability
  • Ventex™ exhalation valve lets the hot air out faster to enable easier breathing (Fisher cat #19-047-826 and 19-047-828 only)
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1

Specifications

Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2003, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N100
Color White
Design Type Cup/Molded
Disposable Single-use
Filter Type N100
Material Dura-Mesh
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve Yes
Filter Efficiency 99%
For Use With (Application) Pharmaceutical or battery manufacturing, welding and metal pouring
Material Headstrap Cloth
Model 2730N100
Nose Piece Material Soft foam
Quantity 5/Pk.
Strap Attachment Type HandyStrap™ with Buckle
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.