Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Moldex™ HandyStrap™ Series N95 Particulate Respirators
SDP

Catalog No. 191301352 Shop All Moldex Metric Inc. Products
Change view
Click to view available options
Size:
Medium/Large
With Exhalation Valve:
No
Yes
Nuisance Odor Relief:
None
Organic Vapors
Color:
Black
White
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size With Exhalation Valve Nuisance Odor Relief Color
19-130-1352 Medium/Large Yes Organic Vapors Black
19-039-694B Medium/Large No None White
19-037-501 Medium/Large Yes None White
19-039-695B Medium/Large Yes Organic Vapors White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 19-130-1352 Supplier Moldex™ Supplier No. M2800N95
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

N95 disposable respirators feature a unique HandyStrap™ with buckle, allowing the mask to hang down around the user’s neck when not in use.

  • Cloth strap is cool and comfortable against the skin
  • HandyStrap™ with buckle, allows mask to hang down around the user's neck when not in use
  • Dura-Mesh™ provides bi-directional protection from dirt on the outside and moisture on the inside
  • Soft foam nose cushion for added comfort and no pressure points
  • Softspun™ lining for increased comfort and durability
  • Ventex™ exhalation valve lets the hot air out faster to enable easier breathing (all models EXCEPT 19-039-694A and -694B)
  • Special Ops™ models (19-160-0164 and 19-130-1352) are designed for military, armed forces, law enforcement and personal use
  • Meets heat and flame resistance in accordance with ANSI/ISEA 110-2003 Section 7.11.1
  • N95 Plus Nuisance Organic Vapor Relief Applications (19-039-695B and 19-130-1352)

Specifications

Aerosol Type N - Not Resistant to Oil
ANSI Standard ANSI/ISEA 110-2009, Section 7.11.1
Certifications (NIOSH) NIOSH 42 CFR 84, N95
Color Black
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Dura-Mesh
Product Type General Purpose Particulate Respirator
Size Medium/Large
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Military, armed forces, federal and local law enforcement, personal use.
Material Headstrap Cloth
Model M2800N95
Nose Piece Material Soft foam
Nosepiece Type Soft foam nose cushion
Nuisance Odor Relief Organic Vapors
Quantity 10/Pk.
Strap Attachment Type HandyStrap™ with Buckle
FDA Cleared Surgical Mask No
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.